Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPS12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542851
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542851 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542851 Supplier Invitrogen™ Supplier No. PA542851
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LVPRLWATCS MATLNQMHRL GPPKRPPRKL GPTEGRPQLK GVVLCTFTRK Sequence homology: Cow: 86%; Dog: 91%; Guinea Pig: 86%; Horse: 79%; Human: 100%; Rat: 86%.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S12P family. The encoded protein is a key component of the ribosomal small subunit and controls the decoding fidelity and susceptibility to aminoglycoside antibiotics. The gene for mitochondrial seryl-tRNA synthetase is located upstream and adjacent to this gene, and both genes are possible candidates for the autosomal dominant deafness gene (DFNA4). Splice variants that differ in the 5' UTR have been found for this gene; all three variants encode the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPS12
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MRPS12
Gene Accession No. O15235
Gene Alias 28S ribosomal protein S12, mitochondrial; AI327385; mitochondrial ribosomal protein S12; Mitochondrial small ribosomal subunit protein uS12m; mitoribosomal protein s12; MPR-S12; Mrps12; MRP-S12; MT-RPS12; ribosomal protein, mitochondrial, S12; RPMS12; RPS12; RPSM12; S12mt; Unknown (protein for MGC:137645)
Gene Symbols MRPS12
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human MRPS12 (aa 20-69).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6183
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.