Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRPS14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562313
Description
Immunogen sequence: YADERLRINS LRKNTILPKI LQDVADEEIA ALPRDSCPVR IRNRCVMTSR PRGVKRRWRL SRIVFRHLAD HGQLSGIQRA TW Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.

Specifications
MRPS14 | |
Polyclonal | |
Unconjugated | |
MRPS14 | |
1810032L21Rik; 28S ribosomal protein S14, mitochondrial; AL022847; DJ262D12.2; HSMRPS14; mitochondrial 28S ribosomal protein S14; mitochondrial ribosomal protein S14; Mitochondrial small ribosomal subunit protein uS14m; MRPS14; MRP-S14; Rpms14; S14mt | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
63931 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60783 | |
MRPS14 | |
Recombinant protein corresponding to Human MRPS14. Recombinant protein control fragment (Product #RP-101231). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction