Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPS14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120370
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120370 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120370 Supplier Invitrogen™ Supplier No. PA5120370
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: BxPC-3, U-251MG, Mouse heart, Mouse kidney, Mouse liver. The target is usually found in the following locations: Mitochondrion. Immunogen sequence: MAAFMLGSLL RTFKQMVPSS ASGQVRSHYV DWRMWRDVKR RKMAYEYADE RLRINSLRKN TILPKILQDV ADEEIAALPR DSCPVRIRNR CVMTSRPRGV KRRWRLSRIV FRHLADHGQL SGIQRATW.

TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPS14
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MRPS14
Gene Accession No. O60783, Q9CR88
Gene Alias 1810032L21Rik; 28S ribosomal protein S14, mitochondrial; AL022847; DJ262D12.2; HSMRPS14; mitochondrial 28S ribosomal protein S14; mitochondrial ribosomal protein S14; Mitochondrial small ribosomal subunit protein uS14m; MRPS14; MRP-S14; Rpms14; S14mt
Gene Symbols MRPS14
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-128 of human MRPS14 (NP_071383.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63931, 64659
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.