Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPS15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566898
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566898 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566898 Supplier Invitrogen™ Supplier No. PA566898
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EQFMKKIVANP EDTRSLEARI IALSVKIRSY EEHLEKHRKD KAHKRYLLMS IDQRKKMLKN LRNTNYDVFE Highest antigen sequence identity to the following orthologs - mouse 77%, rat 76%.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. MRPS15 encodes a 28S subunit protein that belongs to the ribosomal protein S15P family. The encoded protein is more than two times the size of its E. coli counterpart, with the 12S rRNA binding sites conserved. Between human and mouse, the encoded protein is the least conserved among small subunit ribosomal proteins. Pseudogenes corresponding to this gene are found on chromosomes 15q and 19q.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPS15
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRPS15
Gene Accession No. P82914
Gene Alias 1500003E24Rik; 2410002B11Rik; 28S ribosomal protein S15, mitochondrial; DC37; FLJ11564; mitochondrial ribosomal protein S15; Mitochondrial small ribosomal subunit protein uS15m; Mprs15; MPR-S15; Mrps15; MRP-S15; RPMS15; S15mt
Gene Symbols MRPS15
Host Species Rabbit
Immunogen Recombinant Human MRPS15. Recombinant protein control fragment (Product #RP-107547).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64960
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.