Learn More
Invitrogen™ MRPS18A Polyclonal Antibody

Description
MRPS18A Polyclonal Antibody for ELISA, Western Blot
Specifications
Specifications
| Antigen | MRPS18A |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | Mrps18a |
| Gene Accession No. | Q9NVS2 |
| Gene Alias | 1110004O20Rik; 28S ribosomal protein S18-3, mitochondrial; 28S ribosomal protein S18a, mitochondrial; 39S ribosomal protein S18-3, mitochondrial; 39S ribosomal protein S18a, mitochondrial; C79712; HumanS18b; Mitochondrial large ribosomal subunit protein bS18a; Mitochondrial large ribosomal subunit protein mL66; mitochondrial ribosomal protein S18-3; mitochondrial ribosomal protein S18A; MRP-S18-3; MRPS18-3; MRPS18A; MRP-S18-a; S18bmt; S18mt-a |
| Gene Symbols | Mrps18a |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.