Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPS18B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562171
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562171 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562171 Supplier Invitrogen™ Supplier No. PA562171
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASVLNTVLRR LPMLSLFRGS HRVQVPLQTL CTKAPSEEDS LSSVPISPYK DEPWKYLESE EYQERYGSRP VWADYRRNHK GGVPPQRTRK Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 71%.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S18P family. The encoded protein is one of three that has significant sequence similarity to bacterial S18 proteins. The primary sequences of the three human mitochondrial S18 proteins are no more closely related to each other than they are to the prokaryotic S18 proteins. Pseudogenes corresponding to this gene are found on chromosomes 1q and 2q.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPS18B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRPS18B
Gene Accession No. Q9Y676
Gene Alias 2400002C15Rik; 28S ribosomal protein S18-2, mitochondrial; 28S ribosomal protein S18b, mitochondrial; C6orf14; HSPC183; HumanS18a; mitochondrial ribosomal protein S18-2; mitochondrial ribosomal protein S18B; Mitochondrial small ribosomal subunit protein bS18b; Mitochondrial small ribosomal subunit protein mS40; MRP-S18-2; MRPS18-2; MRPS18B; MRP-S18-b; mrps18-b; PTD017; S18amt; S18mt-b
Gene Symbols MRPS18B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MRPS18B. Recombinant protein control fragment (Product #RP-100645).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28973
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.