Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPS2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen MRPS2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP154663
SDP
View Documents
Novus Biologicals
NBP154663
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

MRPS2 Polyclonal specifically detects MRPS2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

MRPS2
Polyclonal
Rabbit
Core ESC Like Genes, Stem Cell Markers
Q9Y399
51116
Synthetic peptides corresponding to MRPS2(mitochondrial ribosomal protein S2) The peptide sequence was selected from the N terminal of MRPS2. Peptide sequence MATSSAALPRILGAGARAPSRWLGFLGKATPRPARPSRRTLGSATALMIR.
Primary
Western Blot
Unconjugated
RUO
Human
28S ribosomal protein S2, mitochondrial, CGI-91, mitochondrial 28S ribosomal protein S2, mitochondrial ribosomal protein S2, MRP-S2, S2mt
MRPS2
IgG
33 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.