Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPS2 Antibody (OTI4D6), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272783AF350
Description
MRPS2 Monoclonal specifically detects MRPS2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
MRPS2 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
MRPS2 | |
Full length recombinant protein of human MRPS2 (NP_057118) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI4D6 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
28S ribosomal protein S2, mitochondrial, CGI-91, mitochondrial 28S ribosomal protein S2, mitochondrial ribosomal protein S2, MRP-S2, S2mt | |
Mouse | |
Affinity Purified | |
Core ESC Like Genes, Stem Cell Markers | |
51116.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction