Learn More
MRPS2 Antibody (OTI4D6), DyLight 650, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP272783C
Catalog No. NBP272783C
Only null left
Add to Cart
Description
Description
MRPS2 Monoclonal specifically detects MRPS2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
Specifications
MRPS2
Monoclonal
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
MRPS2
Full length recombinant protein of human MRPS2 (NP_057118) produced in E.coli.
0.1 mL
Primary
Human
IgG2a
Western Blot, Flow Cytometry, Immunohistochemistry
OTI4D6
Western Blot, Flow Cytometry, Immunohistochemistry
28S ribosomal protein S2, mitochondrial, CGI-91, mitochondrial 28S ribosomal protein S2, mitochondrial ribosomal protein S2, MRP-S2, S2mt
Mouse
Affinity Purified
Core ESC Like Genes, Stem Cell Markers
51116.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.