Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPS2 Antibody (OTI4D6), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272783MFV500
Description
MRPS2 Monoclonal antibody specifically detects MRPS2 in Human samples. It is validated for Flow CytometrySpecifications
MRPS2 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
OTI4D6 | |
Flow Cytometry | |
28S ribosomal protein S2, mitochondrial, CGI-91, mitochondrial 28S ribosomal protein S2, mitochondrial ribosomal protein S2, MRP-S2, S2mt | |
Full length human recombinant protein of human MRPS2 (NP_057118) produced in E.coli. | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
51116 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction