Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPS21 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298522AF532
Description
MRPS21 Polyclonal specifically detects MRPS21 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
MRPS21 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
28S ribosomal protein S21, mitochondrial, mitochondrial 28S ribosomal protein S21, mitochondrial ribosomal protein S21, MRP-S21, RPMS21, S21mt | |
Produced in rabbits immunized with E. coli-derived Human MRPS21 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
54460 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction