Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPS22 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | MRPS22 |
---|---|
Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
MRPS22 Polyclonal specifically detects MRPS22 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MRPS22 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
56945 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:RMIQVYFPKEGRKILTPIIFKEENLRTMYSQDRHVDVLNLCFAQFEPDSTEYIKVHHKTYEDIDKRGKYDLLRSTRYFGGMVWYFVNNKKIDGLLIDQIQRDLIDDATNLVQLYHVLHPDGQSAQGAKD | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Rabbit | |
Human | |
C3orf5GK002, COXPD5, GIBT, mitochondrial ribosomal protein S22, MRP-S2228S ribosomal protein S22, mitochondrial, RPMS22, S22mt | |
MRPS22 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title