Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRPS22 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551014
Description
MRPS22 Polyclonal Antibody detects endogenous levels of MRPS22 protein.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that does not seem to have a counterpart in prokaryotic and fungal-mitochondrial ribosomes. This gene lies telomeric of and is transcribed in the opposite direction from the forkhead box L2 gene. A pseudogene corresponding to this gene is found on chromosome Xq.
Specifications
MRPS22 | |
Polyclonal | |
Unconjugated | |
MRPS22 | |
28S ribosomal protein S22, mitochondrial; 3100002P07Rik; C3orf5; COXPD5; GIBT; GK002; mitochondrial ribosomal protein S22; Mitochondrial small ribosomal subunit protein mS22; MRPS22; MRP-S22; ribosomal protein, mitochondrial, S22; Rpms22; S22mt | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56945 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P82650 | |
MRPS22 | |
Synthesized peptide derived from the Internal region of human MRP-S22 (aa 231-280). | |
100 μL | |
Primary | |
Human, Monkey | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction