Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPS23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582834
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582834 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582834 Supplier Invitrogen™ Supplier No. PA582834
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FNPNFKSTCQ RFVEKYTELQ KLGETDEEKL FVETGKALLA EGVILRRVGE ARTQHGGSHV SRKSEHLSVR PQTALEENET QKEVPQD Highest antigen sequence indentity to the following orthologs: Mouse - 66%, Rat - 64%.

PHTNS interacts with regulatory subunits that target the enzyme to different cellular locations and change its activity toward specific substrates. Phostensin is a regulatory subunit that targets PP1 to F-actin cytoskeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPS23
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRPS23
Gene Accession No. Q9Y3D9
Gene Alias 28S ribosomal protein S23, mitochondrial; CGI-138; D11Bwg1153e; HSPC329; mitochondrial ribosomal protein S23; Mitochondrial small ribosomal subunit protein mS23; MRPS23; MRP-S23; Rpms23; S23mt
Gene Symbols MRPS23
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MRPS23. Recombinant protein control fragment (Product #RP-93498).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51649
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.