Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRPS24 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554295
Description
Immunogen sequence: GTFPGCLADQ LVLKRRGNQL EICAVVLRQL SPHKYYFLVG YSETLLSYFY KCPVRLHLQT VPSKVVYKY Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. MRPS24 encodes a 28S subunit protein. A pseudogene corresponding to this gene is found on chromosome 11.
Specifications
MRPS24 | |
Polyclonal | |
Unconjugated | |
MRPS24 | |
28S ribosomal protein S24, mitochondrial; 3110030K20Rik; AI414579; bMRP47; bMRP-47; DD6A4-3; HSPC335; mitochondrial 28S ribosomal protein S24; mitochondrial ribosomal protein S24; Mitochondrial small ribosomal subunit protein uS3m; MRPS24; MRP-S24; ribosomal protein, mitochondrial, S24; Rpms24; S24mt | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
498406, 64951 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96EL2 | |
MRPS24 | |
Recombinant protein corresponding to Human MRPS24. Recombinant protein control fragment (Product #RP-92621). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction