Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPS26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531595
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531595 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531595 Supplier Invitrogen™ Supplier No. PA531595
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Raji. Predicted reactivity: Rhesus Monkey (91%), Bovine (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein. This gene lies adjacent to and downstream of the gonadotropin-releasing hormone precursor gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPS26
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene MRPS26
Gene Accession No. Q9BYN8
Gene Alias 28S ribosomal protein S13, mitochondrial; 28S ribosomal protein S26, mitochondrial; 5'OT-EST protein; AI648866; C20orf193; dJ534B8.3; GI008; mitochondrial ribosomal protein S26; Mitochondrial small ribosomal subunit protein mS26; MRPS13; MRP-S13; Mrps26; MRP-S26; NY-BR-87; protein 5'OT-EST; RPMS13; rt26; S13mt; S26mt; serologically defined breast cancer antigen NY-BR-87
Gene Symbols MRPS26
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human MRPS26. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64949
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.