Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPS27 Antibody (OTI6E5), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272787PE
Description
MRPS27 Monoclonal specifically detects MRPS27 in Human samples. It is validated for Flow Cytometry.Specifications
MRPS27 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
MRPS27 | |
Recombinant protein fragment corresponding to amino acids 151-414 of human MRPS27 (NP_055899) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Flow Cytometry | |
OTI6E5 | |
Flow Cytometry | |
FLJ21764, KIAA0264FLJ23348, mitochondrial, mitochondrial ribosomal protein S27, MRP-S27 | |
Mouse | |
Affinity Purified | |
Stem Cell Markers | |
23107.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction