Learn More
Invitrogen™ MRPS30 Polyclonal Antibody

Description
The antibody detects endogenous levels of total MRPS30 protein.
Specifications
Specifications
| Antigen | MRPS30 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Mrps30 |
| Gene Accession No. | Q9NP92 |
| Gene Alias | 2610020A16Rik; 28S ribosomal protein S30, mitochondrial; 39S ribosomal protein S30, mitochondrial; AA968347; BM-047; Mitochondrial large ribosomal subunit protein mL65; Mitochondrial large ribosomal subunit protein mS30; mitochondrial ribosomal protein S30; MRP S30; Mrps30; MRP-S30; PAP; PDCD9; programmed cell death 9; Programmed cell death protein 9; S30mt |
| Gene Symbols | Mrps30 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.