Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPS31 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23055925UL
Description
MRPS31 Polyclonal specifically detects MRPS31 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MRPS31 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Q92665 | |
MRPS31 | |
This antibody was developed against a recombinant protein corresponding to amino acids: QLATVNEQPLQNGFEELIQWTKEGKLWEFPINNEAGFDDDGSEFHEHIFLEKHLESFPKQGPIRHFMELVTCGLSKNPYLSVKQKVEH | |
25ul | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
28S ribosomal protein S31, mitochondrial, IMOGN38Imogen 38, mitochondrial ribosomal protein S31, MRP-S31, S31mt | |
Rabbit | |
Affinity Purified | |
RUO | |
10240 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction