Learn More
Invitrogen™ MRPS34 Monoclonal Antibody (OTI4C9)

Description
MRPS34 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | MRPS34 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI4C9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MRPS34 |
| Gene Accession No. | P82930 |
| Gene Alias | 0610007F04Rik; 28S ribosomal protein S34, mitochondrial; 5330430D13Rik; AV001970; mitochondrial 28S ribosomal protein S34; mitochondrial ribosomal protein S34; Mitochondrial small ribosomal subunit protein mS34; MRPS12; MRP-S12; MRPS34; MRP-S34; rt34; S34mt; Tce2; T-complex expressed gene 2 protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.