Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPS34 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559872
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559872 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559872 Supplier Invitrogen™ Supplier No. PA559872
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PEDSLASVPY PPLLRAMIIA ERQKNGDTST EEPMLNVQRI RMEPWDYPAK QEDKGRAKGT Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 80%.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein. Alternate splice variants of this gene have been described but their full-length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPS34
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRPS34
Gene Accession No. P82930, Q9JIK9
Gene Alias 0610007F04Rik; 28S ribosomal protein S34, mitochondrial; 5330430D13Rik; AV001970; mitochondrial 28S ribosomal protein S34; mitochondrial ribosomal protein S34; Mitochondrial small ribosomal subunit protein mS34; MRPS12; MRP-S12; MRPS34; MRP-S34; rt34; S34mt; Tce2; T-complex expressed gene 2 protein
Gene Symbols MRPS34
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MRPS34. Recombinant protein control fragment (Product #RP-98609).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287126, 65993, 79044
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.