Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPS35 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558421
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558421 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558421 Supplier Invitrogen™ Supplier No. PA558421
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DMEEYIWENS SSERNILETL LQMKAAEKNM EINKEELLGT KEIEEYKKSV VSLKNEEENE NSISQYKESV K Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 65%.

Mammalian mitochondrial ribosomes (mitoribosomes) are responsible for protein synthesis within the mitochondrion. The mitoribosomes are composed of a 4:1 ratio of protein to RNA, with the proteins forming two subunits, the 28S subunit and the 39S subunit. Across species, the proteins that make up the mitoribosome subunits vary greatly in sequence, preventing easy recognition by sequence homology. MRP-S35 (mitochondrial ribosomal protein S35), also known as MDS023, MRPS28 or HDCMD11P, is a 323 amino acid protein that localizes to the mitochondrion, where it exists as a component of the 28S ribosomal subunit and works in conjunction with other MRPs to mediate protein synthesis. Existing as two alternatively spliced isoforms, MRP-S35 is encoded by a gene located on human chromosome 10, which houses over 1,200 genes and comprises nearly 4. 5% of the human genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPS35
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRPS35
Gene Accession No. P82673
Gene Alias 28S ribosomal protein S28, mitochondrial; 28S ribosomal protein S35, mitochondrial; HDCMD11P; MDS023; MDSO23; mitochondrial ribosomal protein S28; mitochondrial ribosomal protein S34; mitochondrial ribosomal protein S35; Mitochondrial small ribosomal subunit protein mS35; MRPS28; MRP-S28; Mrps35; MRP-S35; PSEC0213; S28mt; S35mt
Gene Symbols MRPS35
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MRPS35. Recombinant protein control fragment (Product #RP-97649).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60488
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.