Learn More
Invitrogen™ MRPS36 Polyclonal Antibody

Description
The antibody detects endogenous levels of total MRPS36 protein.
Specifications
Specifications
| Antigen | MRPS36 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | MRPS36 |
| Gene Accession No. | P82909 |
| Gene Alias | 1110018B13Rik; 28S ribosomal protein S36, mitochondrial; DC47; mitochondrial ribosomal protein S36; Mrps36; MRP-S36; RT36; S36mt |
| Gene Symbols | MRPS36 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.