Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPS36 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575468
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575468 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575468 Supplier Thermo Scientific Supplier No. PA575468
Add to Cart
Add to Cart

MRPS36 Polyclonal Antibody

MRPS36 Polyclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPS36
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene MRPS36
Gene Accession No. P82909, Q9CQX8
Gene Alias 1110018B13Rik; 28S ribosomal protein S36, mitochondrial; DC47; mitochondrial ribosomal protein S36; Mrps36; MRP-S36; RT36; S36mt
Gene Symbols MRPS36
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the N-terminal of Human MRPS36.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 294696, 66128, 92259
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.