Learn More
Invitrogen™ MRPS5 Polyclonal Antibody

Description
The antibody detects endogenous levels of total MRPS5 protein.
Specifications
Specifications
| Antigen | MRPS5 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | MRPS5 |
| Gene Accession No. | P82675 |
| Gene Alias | 1620401I16Rik; 28S ribosomal protein S5, mitochondrial; AI850294; mitochondrial 28S ribosomal protein S5; mitochondrial ribosomal protein S5; Mitochondrial small ribosomal subunit protein uS5m; MRPS5; MRP-S5; S5mt |
| Gene Symbols | MRPS5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.