Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPS6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576449
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576449 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576449 Supplier Thermo Scientific Supplier No. PA576449
Add to Cart
Add to Cart

MRPS6 Polyclonal Antibody

MRPS6 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPS6
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene MRPS6
Gene Accession No. P58064, P82932
Gene Alias 28S ribosomal protein S6, mitochondrial; AW046321; C21orf101; LOC100360017; mitochondrial ribosomal protein S6; mitochondrial ribosomal protein S6-like; Mitochondrial small ribosomal subunit protein bS6m; MRPS6; MRP-S6; Rpms6; S6mt
Gene Symbols MRPS6
Host Species Rabbit
Immunogen Recombinant protein of human MRPS6.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100360017, 121022, 64968
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.