Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPS6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552475
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552475 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552475 Supplier Invitrogen™ Supplier No. PA552475
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KQPETAATLK RTIEALMDRG AIVRDLENLG ERALPYRISA HSQQHNRGGY FLVDFYAPTA AVESMVEHLS RDIDVIRGNI VKHPLTQELK ECEGIVPVPL AEKLYSTKKR KK Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 85%.

Mitochondrial ribosomes are made of a 28S subunit and a larger 39S subunit. These ribosomes have an approximate composition of 75% protein to rRNA as compared to prokaryotic ribosomes, where reverse proportions are found. MRP-S6 (Mitochondrial 28S ribosomal protein S6) is a mitochondrial ribosomal protein that belongs to the ribosomal protein S6P family. MRP-S6 is a component of the mitochondrial ribosome small subunit (28S) which is comprised of a 12S rRNA and almost 30 other distinct proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPS6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRPS6
Gene Accession No. P82932
Gene Alias 28S ribosomal protein S6, mitochondrial; AW046321; C21orf101; LOC100360017; mitochondrial ribosomal protein S6; mitochondrial ribosomal protein S6-like; Mitochondrial small ribosomal subunit protein bS6m; MRPS6; MRP-S6; Rpms6; S6mt
Gene Symbols MRPS6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MRPS6. Recombinant protein control fragment (Product #RP-89873).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64968
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.