Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPS7 Antibody (OTI5E11), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272789AF405
Description
MRPS7 Monoclonal specifically detects MRPS7 in Human samples. It is validated for Western Blot.Specifications
MRPS7 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MRPS7 | |
Recombinant protein fragment corresponding to amino acids 38-242 of human MRPS7 (NP_057055) produced in E.coli. | |
0.1 mL | |
51081.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI5E11 | |
Western Blot | |
bMRP27a, bMRP-27a, mitochondrial ribosomal protein S7, MRP-S, MRP-S7,30S ribosomal protein S7 homolog, RPMS7,28S ribosomal protein S7, mitochondrial, RP-S7, S7mt | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction