Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPS7 Antibody (OTI5E11), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245433
Description
Ensure accurate, reproducible results in Western Blot
MRPS7 Monoclonal specifically detects MRPS7 in Human samples. It is validated for Western Blot.Specifications
MRPS7 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000 | |
bMRP27a, bMRP-27a, mitochondrial ribosomal protein S7, MRP-S, MRP-S7,30S ribosomal protein S7 homolog, RPMS7,28S ribosomal protein S7, mitochondrial, RP-S7, S7mt | |
Mouse | |
28 kDa | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
OTI5E11 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
MRPS7 | |
Human recombinant protein fragment corresponding to amino acids 38-242 of human MRPS7 (NP_057055) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
51081 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction