Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPS9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

Supplier:  Proteintech Group Inc 165331AP150UL

Encompass_Preferred

 View more versions of this product

Catalog No. 50-173-1012


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

Mammalian mitochondrial ribosomes (mitoribosomes) are responsible for protein synthesis within the mitochondrion. The mitoribosomes are composed of a 4:1 ratio of protein to RNA, with the proteins forming two subunits, the 28S subunit and the 39S subunit. Across species, the proteins that make up the mitoribosome subunits vary greatly in sequence, preventing easy recognition by sequence homology. MRP-S9 (mitochondrial 28S ribosomal protein S9), also known as RPMS9, is a 396 amino acid mitochondrial ribosomal protein. Localized to mitochondria, MRP-S9 is present in the 28S subunit of the mitoribosomes, which comprises a 12S rRNA and at least 30 distinct proteins.
Specifications

Specifications

MRPS9
Polyclonal
Unconjugated
MRPS9
MRP S9, MRPS9, RPMS9, S9mt
Rabbit
Antigen Affinity Chromatography
RUO
301371, 64965, 69527
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P82933, Q9D7N3
MRPS9
MRPS9 Fusion Protein Ag9809
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.