Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRRF Antibody (OTI1D5), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272790V
Description
MRRF Monoclonal specifically detects MRRF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MRRF | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MRRF | |
Recombinant protein fragment corresponding to amino acids 56-262 of human MRRF (NP_620132) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1D5 | |
Western Blot, Immunohistochemistry | |
mitochondrial, mitochondrial ribosome recycling factor, MTRRF | |
Mouse | |
Affinity Purified | |
DNA replication Transcription Translation and Splicing | |
92399.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction