Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRRF Monoclonal Antibody (OTI2A9)
GREENER_CHOICE

Catalog No. PIMA526777
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526777 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526777 Supplier Invitrogen™ Supplier No. MA526777
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MRRF Monoclonal Antibody for Western Blot, IHC (P)

Responsible for the release of ribosomes from messenger RNA at the termination of protein biosynthesis. May increase the efficiency of translation by recycling ribosomes from one round of translation to another. Ota T., Nat. Genet. 36:40-45(2004). Humphray S.J., Nature 429:369-374(2004). Zhang Y., Biochim. Biophys. Acta 1443:245-250(1998).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRRF
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2A9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Mrrf
Gene Accession No. Q96E11
Gene Alias 2400002D02Rik; mitochondrial ribosome recycling factor; MRFF; MRRF; MTRRF; Ribosome-recycling factor, mitochondrial; ribosome-releasing factor, mitochondrial; RRF
Gene Symbols Mrrf
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 56-262 of MRRF produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92399
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.