Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ MRS 1754
GREENER_CHOICESDP

Catalog No. 275210 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-521-0 10 mg
27-525-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 27-521-0 Supplier R&D Systems™ Supplier No. 2752/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective A2B antagonist

Selective adenosine A2B receptor antagonist (Ki values are 1.97, 16.8, 403, 503, 570 and 612 nM for hA2B, rA1, hA1, hA2A, hA3 and rA2A receptors respectively).

Chemical Identifiers

CAS 264622-58-4
Molecular Formula C26H26N6O4

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 486.52
Purity >98%
Molecular Weight (g/mol) 486.532
Chemical Name or Material MRS 1754
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.