Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MRS 1754

Catalog No. 275250
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-525-0 50 mg
27-521-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 27-525-0 Supplier Tocris Bioscience™ Supplier No. 2752/50
This item is not returnable. View return policy

Selective A2B antagonist

Selective adenosine A2B receptor antagonist (Ki values are 1.97, 16.8, 403, 503, 570 and 612 nM for hA2B, rA1, hA1, hA2A, hA3 and rA2A receptors respectively).

Chemical Identifiers

CAS 264622-58-4
Molecular Formula C26H26N6O4

Specifications

Quantity 50 mg
Formula Weight 486.52
Purity >98%
Molecular Weight (g/mol) 486.532
Chemical Name or Material MRS 1754
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.