Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MRS 2690
GREENER_CHOICESDP

Catalog No. 29151 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-151 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 29-151 Supplier Tocris Bioscience™ Supplier No. 2915/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent P2Y14 agonist

MRS 2690 is a potent P2Y14 receptor agonist (EC50 = 49 nM) that displays 7-fold higher potency than UDP-glucose. This product is typically reconstituted in water.

Chemical Identifiers

CAS 15039-58-4
Molecular Formula C15H22N2Na2O16P2S

Specifications

Quantity 1 mg
Formula Weight 626.33
Purity >98%
Chemical Name or Material MRS 2690
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.