Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ MRS 2693 trisodium salt
GREENER_CHOICESDP

Catalog No. 25021 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-021 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 25-021 Supplier R&D Systems™ Supplier No. 2502/1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective P2Y6 agonist

Selective P2Y6 agonist (EC50 = 0.015 μM at the hP2Y6 receptor). Displays no activity at other P2Y subtypes. Exerts a cytoprotective effect in an in vivo mouse model of hindlimb skeletal muscle ischemia/reperfusion injury.

Chemical Identifiers

CAS 1448858-83-0
Molecular Formula C9H13IN2Na3O12P2

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 596
Purity >95%
Molecular Weight (g/mol) 599.026
Chemical Name or Material MRS 2693 trisodium salt
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.