Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MRS 2768 tetrasodium salt
GREENER_CHOICESDP

Catalog No. 38841 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-841 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 38-841 Supplier Tocris Bioscience™ Supplier No. 3884/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective P2Y2 agonist

Selective P2Y2 agonist (EC50 = 1.89 μM). Displays no affinity for human P2Y4 or P2Y6 receptors.

Chemical Identifiers

CAS 2567869-47-8
Molecular Formula C15H16N2Na4O18P4

Specifications

Quantity 1 mg
Formula Weight 728.14
Purity >98%
Molecular Weight (g/mol) 728.143
Chemical Name or Material MRS 2768 tetrasodium salt
Recommended Storage Store at -80°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.