Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                MRS2 Antibody, Novus Biologicals™
 
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
| Antigen | MRS2 | 
|---|---|
| Applications | Western Blot | 
| Classification | Polyclonal | 
| Conjugate | Unconjugated | 
| Host Species | Rabbit | 
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
| NBP15961420  | Novus Biologicals NBP15961420UL | 20 μL | 
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $206.00
                                                 |  | |||||
| NBP159614  | Novus Biologicals NBP159614 | 100 μL | 
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $487.50
                                                 |  | |||||
Description
MRS2 Polyclonal specifically detects MRS2 in Human samples. It is validated for Western Blot.Specifications
| MRS2 | |
| Polyclonal | |
| Rabbit | |
| Q9HD23 | |
| 57380 | |
| Synthetic peptides corresponding to MRS2L The peptide sequence was selected from the middle region of MRS2L. Peptide sequence LDALVDPKHSSVDRSKLHILLQNGKSLSELETDIKIFKESILEILDEEEL. | |
| Primary | 
| Western Blot | |
| Unconjugated | |
| RUO | |
| HPT, MGC78523, mitochondrial, MRS2 magnesium homeostasis factor homolog (S. cerevisiae), MRS2-like protein, MRS2-like, magnesium homeostasis factor, MRS2-like, magnesium homeostasis factor (S. cerevisiae) | |
| MRS2 | |
| IgG | |
| 50 kDa | 
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            