Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553616
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553616 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553616 Supplier Invitrogen™ Supplier No. PA553616
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NTLQGKLSIL QPLILETLDA LVDPKHSSVD RSKLHILLQN GKSLSELETD IKIFKESILE ILDEEELLEE LCVSKWSDPQ VFEKSSAGID HAEEMELLLE NYYRLADDLS NAARELRVLI DDSQSIIFI Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 93%.

Mrs2 is a magnesium transporter that mediates the influx of magnesium into the mitochondrial matrix. Mrs2 is required for normal expression of the mitochondrial respiratory complex I subunits, and mediates the influx of magnesium into the mitochondrial matrix. Mrs2 is also required for normal expression of the mitochondrial respiratory complex I subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRS2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRS2
Gene Accession No. Q9HD23
Gene Alias Gm902; HPT; Magnesium transporter MRS2 homolog, mitochondrial; MRS2; MRS2 magnesium homeostasis factor homolog; MRS2 magnesium homeostasis factor homolog (S. cerevisiae); MRS2 magnesium transporter; MRS2, magnesium transporter; MRS2L; MRS2-like protein; MRS2-like, magnesium homeostasis factor; putative magnesium transporter; RPT; RPT protein similar to yeast MRS2
Gene Symbols MRS2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MRS2. Recombinant protein control fragment (Product #RP-90946).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57380
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.