Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ MRT 67307 dihydrochloride
GREENER_CHOICESDP

Catalog No. 513410 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-341-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 51-341-0 Supplier R&D Systems™ Supplier No. 5134/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Salt inducible kinase (SIK) inhibitor

MRT 67307 dihydrochloride is a salt inducible kinase (SIK) inhibitor (IC50 values are 67, 250 and 430 nM for SIK2, SIK1 and SIK3 respectively). Also inhibits TBK1, MARK1-4, IKKε and NUAK1 (IC50 values are 19, 27-52, 160 and 230 nM respectively). Has no effect on IKKα or IKKβ. Induces IL-10 secretion and inhibits TNF-α and IL-6 secretion in bacterial LPS-stimulated macrophages. Also enhances IL-1-induced activation of NFκB-dependent gene transcription in mouse embryonic fibroblast (MEF) cells. Inhibits autophagy.

Specifications

CAS 1781882-89-0
Quantity 10 mg
Product Line Tocris
Product Type MRT 67307 dihydrochloride
Purity >98%
Molecular Weight (g/mol) 537.52
Inhibitors MRT 67307 dihydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.