Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRTO4 Antibody, Novus Biologicals™
SDP

Catalog No. p-7106380 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP153203 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP153203 Supplier Novus Biologicals Supplier No. NBP153203
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MRTO4 Polyclonal specifically detects MRTO4 in Human samples. It is validated for Western Blot.

Specifications

Antigen MRTO4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9UKD2
Gene Alias C1orf33, chromosome 1 open reading frame 33, dJ657E11.4, mRNA turnover 4 homolog (S. cerevisiae), mRNA turnover protein 4 homolog, MRT4, mRNA turnover 4, homolog, MRT4, mRNA turnover 4, homolog (S. cerevisiae), MRT460S acidic ribosomal protein PO
Gene Symbols MRTO4
Host Species Rabbit
Immunogen Synthetic peptides corresponding to MRTO4(mRNA turnover 4 homolog (S. cerevisiae)) The peptide sequence was selected from the N terminal of MRTO4. Peptide sequence SKLKDIRNAWKHSRMFFGKNKVMMVALGRSPSDEYKDNLHQVSKRLRGEV.
Molecular Weight of Antigen 27 kDa
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 51154
Test Specificity Zebrafish: 77%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.