Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRTO4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542839
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542839 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542839 Supplier Thermo Scientific Supplier No. PA542839
Add to Cart
Add to Cart

MRTO4 Polyclonal Antibody

MRTO4 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen MRTO4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MRTO4
Gene Accession No. Q9UKD2
Gene Alias 2610012O22Rik; 60S acidic ribosomal protein PO; C1orf33; dJ657E11.4; fb71a02; hypothetical protein LOC550350; Mg684; mRNA turnover 4 homolog; mRNA turnover 4 homolog (S. cerevisiae); mRNA turnover protein 4 homolog; MRT4; MRT4 homolog, ribosome maturation factor; MRT4, mRNA turnover 4, homolog; MRT4, mRNA turnover 4, homolog (S. cerevisiae); mrto4; MRTO4 ribosome maturation factor; muscle protein684; RGD1311709; Ribosome assembly factor MRTO4; wu:fb71a02; zgc:110388
Gene Symbols MRTO4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human MRTO4.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51154
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.