Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRTO4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555313
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555313 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555313 Supplier Invitrogen™ Supplier No. PA555313
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YKDNLHQVSK RLRGEVGLLF TNRTKEEVNE WFTKYTEMDY ARAGNKAAFT VSLDPGPLEQ FPHSMEPQLR QLGLPTALKR GVV Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

MRTO4 is a protein sharing a low level of sequence similarity with ribosomal protein P0. While the precise function of the protein is currently unknown, it appears to be involved in mRNA turnover and ribosome assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRTO4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRTO4
Gene Accession No. Q9UKD2
Gene Alias 2610012O22Rik; 60S acidic ribosomal protein PO; C1orf33; dJ657E11.4; fb71a02; hypothetical protein LOC550350; Mg684; mRNA turnover 4 homolog; mRNA turnover 4 homolog (S. cerevisiae); mRNA turnover protein 4 homolog; MRT4; MRT4 homolog, ribosome maturation factor; MRT4, mRNA turnover 4, homolog; MRT4, mRNA turnover 4, homolog (S. cerevisiae); mrto4; MRTO4 ribosome maturation factor; muscle protein684; RGD1311709; Ribosome assembly factor MRTO4; wu:fb71a02; zgc:110388
Gene Symbols MRTO4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MRTO4. Recombinant protein control fragment (Product #RP-93992).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51154
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.