Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MRTX 849
GREENER_CHOICESDP

Catalog No. 748850 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-885-0 50 mg
74-881-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 74-885-0 Supplier Tocris Bioscience™ Supplier No. 7488/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mutation-selective inhibitor of KRASG12C

MRTX 849 is a mutant-selective inhibitor of KRASG12C (IC50 = 142 nM). MRTX 849 covalently binds to KRASG12C at the cysteine at residue 12 and inhibits KRAS-dependent signal transduction by locking the protein in its inactive GDP-bound state. MRTX 849 induces pronounced tumor regression in 17 of 26 (65%) KRASG12C -positive cell lines and patient-derived xenograft models from multiple tumor types. It is orally bioavailable.

Specifications

Chemical Name or Material 4-[7-(8-Chloro-1-naphthalenyl)-5,6,7,8-tetrahydro-2-[[(2 S)-1-methyl-2-pyrrolidinyl]methoxy]pyrido[3,4-d]pyrimidin-4-yl]-1-(2-fluoro-1-oxo-2-propen-1-yl)-(2 S)-2-piperazineacetonitrile
CAS 2326521-71-3
Quantity 50 mg
Target Ras GTPase Inhibitors
Molecular Formula C32H35ClFN7O2
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.