Learn More
Invitrogen™ MRVI1 Polyclonal Antibody

Description
PA3-851 detects CARP from mouse and bovine samples. PA3-851 has been successfully used in Western blot and immunocytochemistry procedures. The PA3-851 immunogen is a synthetic peptide corresponding to residues 93-107 of bovine IRAG.
Specifications
Specifications
| Antigen | MRVI1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Whole serum with 0.05% sodium azide |
| Gene | MRVI1 |
| Gene Accession No. | Q9N1F0, Q9WUX5 |
| Gene Alias | BB115629; inositol 1,4,5-triphosphate receptor associated 1; inositol 1,4,5-triphosphate receptor associated 1; protein MRVI1; inositol 1,4,5-triphosphate receptor-associated cGMP kinase substrate; inositol 1,4,5-trisphosphate receptor-associated cGMP kinase substrate; IP3 receptor associated cGMP kinase substrate; IP3 receptor associated cGMP kinase substrate A; IP3 receptor associated cGMP kinase substrate B; IP3R-associated cGMP kinase substrate; IRAG; IRAG1; JAW1L; JAW1-related protein MRVI1; MRV integration site 1; MRVI1; murine retrovirus integration site 1 homolog; murine retrovirus integration site 1 protein; Protein MRVI1; retroviral integration site 1; Ris1 |
| Gene Symbols | MRVI1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.