Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MS 023 dihydrochloride
GREENER_CHOICE

Catalog No. p-7110882
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-131-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 57-131-0 Supplier Tocris Bioscience™ Supplier No. 5713/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective type I PRMT inhibitor

MS 023 dihydrochloride is a potent and selective type I PRMT inhibitor (IC50 values are 8, 8, 30, 83 and 119 nM for PRMT6, PRMT8, PRMT1, PRMT4 and PRMT3, respectively). Exhibits no significant activity against type II and III PRMTs, PKMTs, DNMTs, KDMs and reader proteins. Reduces global arginine asymmetric dimethylation levels in vitro.

Specifications

Chemical Name or Material N1-Methyl-N1-[[4-[4-(1-methylethoxy)phenyl]-1H-pyrrol-3-yl]methyl]-1,2-ethanediamine dihydrochloride
CAS 1992047-64-9
Quantity 10 mg
Target Histone Methyltransferase Inhibitors
Molecular Formula C17H25N3O.2HCl
Formula Weight 360.32
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.