Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MS 4322
SDP

Catalog No. 80802 Shop All R&D Systems Products
Change view
Click to view available options
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
80-802 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 80-802 Supplier Tocris Bioscience™ Supplier No. 8080/2
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective PRMT5 Degrader (PROTAC™)

MS 4322 is a protein arginine methyltransferase 5 (PRMT5) Degrader (PROTAC™) (DC50 = 1.1 μM, Dmax = 74% in MCF-7 cells). Also inhibits PRMT5 methyltransferase activity (IC50 = 18 nM) and dose-dependently inhibits proliferation of HeLa, A549, A172 and Jurkat cancer cell lines. Comprises a PRMT5 inhibitor linked to an E3 ligase ligand.

PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.

Specifications

Chemical Name or Material N-((S)-3-(3,4-Dihydroisoquinolin-2(1H)-yl)-2-hydroxypropyl)-6-((1-((S)-19-((2S,4R)-4-hydroxy-2-(((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)carbamoyl)pyrrolidine-1-carbonyl)-20,20-dimethyl-17-oxo-3,6,9,12,15-pentaoxa-18-azahenicosanoyl)azetidin-3-yl)amin
CAS 2375432-47-4
Quantity 2 mg
Target Active Degraders
Molecular Formula C55H76N10O12S
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.