Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ MS 436
Potent and selective BRD4(1) inhibitor
Specifications
Purity | >98% |
---|---|
Product Type | MS 436 |
Molecular Weight (g/mol) | 383.42 |
Inhibitors | MS 436 |
Description
Potent and selective BRD4 bromodomain inhibitor (Ki = 30 - 50 nM for the first bromodomain (BRD4(1)). Exhibits 10-fold selectivity for BRD4(1) over BRD4(2). Blocks BRD4 transcriptional activity in lipopolysaccharide-induced production of both nitric oxide and IL-6 in mouse macrophages (IC50 values are 3.8 and 4.9 μM, respectively). Attenuates melanoma cell proliferation in vitro.Specifications
>98% | |
MS 436 | |
383.42 | |
MS 436 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title