Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD103 Rat anti-Mouse, APC-R700, Clone: M290, BD

Rat Monoclonal Antibody

Supplier:  BD Biosciences 565529

Catalog No. BDB565529


Only null left
Add to Cart

Description

Description

The M290 antibody specifically binds to CD103, the α chain of α IEL β 7 integrin. CD103 has a unique and fairly restricted tissue distribution. It is expressed on almost all intestinal intraepithelial lymphocytes (IEL), on dendritic epidermal T cells (DEC), on subpopulations of peripheral T cells, and on distinct subsets of fetal, neonatal, and adult thymocytes. E-cadherin is the epithelial-cell ligand for α IEL β 7 integrin. The ordered expression of α IEL during thymocyte development (which occurs under the influence of the thymic epithelium), the high level of expression of α α IEL β 7-mediated adhesion in in vitro assays.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD103
Monoclonal
0.2mg/mL
Itgae; Integrin alpha-E; αE; alpha-E1; ITAE; Integrin αIEL chain; aM290
Rat
Affinity Purified
RUO
Mouse
IgG2a κ
Flow Cytometry
M290
APC-R700
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Mouse Intestinal Epithelial Cells
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.