Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD66a (CEACAM1a) Mouse anti-Mouse , PE, Clone: CC1 (also known as MAb-CCl; mAb CC1), BD Biosciences™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566828
Description
CD66a (CEACAM1a) Monoclonal antibody specifically detects CD66a (CEACAM1a) in Mouse samples. It is validated for Flow Cytometry
Specifications
CD66a (CEACAM1a) | |
Monoclonal | |
PE | |
CEACAM1[a]; Ceacam1; BGP-1; Bgp; MHV-R; MHVR | |
BALB/c mouse intestinal brush border membrane proteins | |
RUO | |
Mouse | |
IgG1 κ |
Flow Cytometry | |
CC1 (also known as MAb-CCl; mAb CC1) | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction